missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Apoptosis Activator VII, Apoptozole

Catalog No. 17849710MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17849710MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17849710MG Supplier MilliporeSigma™ Supplier No. 17849710MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Apoptosis Activator VII, Apoptozole, also referenced under CAS 1054543-47-3, modulates Apoptosis. This small molecule/inhibitor is primarily used for Cancer applications.

Packaged Under Inert Gas

Specifications

Color White
Description 95% by HPLC
CAS 1054543-47-3
Molecular Formula C33H24F6N3O3
Formula Weight 625.6g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 10 mg