missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Aprotinin, Bovine Lung, Crystalline

Aprotinin, Bovine Lung, Crystalline, CAS 9087-70-1, is a competitive, reversible, heat- and acid-stable inhibitor of proteolytic and esterolytic activities.

$122.38 - $815.05

Specifications

Concentration 5500 KIU/mg, dry weight
Color White to Off-white
Target Proteins chymotrypsin
CAS 9087-70-1
Formula Weight 6512g/mol
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
61637010MG
View Documents
MilliporeSigma™
61637010MG
10 mg
Each for $122.38
Only null left
 
61-637-020MG
View Documents
MilliporeSigma™
61637020MG
20 mg
Each for $213.62
Only null left
 
61-637-0100MG
View Documents
MilliporeSigma™
616370100MG
100 mg
Each for $815.05
Only null left
 
Description

Description

Contaminants: endotoxin: ≤10 EU/mg
Specifications

Specifications

5500 KIU/mg, dry weight
chymotrypsin
6512g/mol
Hygroscopic
95% by gel filtration
White to Off-white
9087-70-1
H2O
Crystalline Solid
Videos
SDS
Documents

Documents