missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Aprotinin, Bovine Lung, Crystalline

Catalog No. 61637020MG
Change view
Click to view available options
Quantity:
10 mg
20 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61637020MG 20 mg
61637010MG 10 mg
616370100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 61637020MG Supplier MilliporeSigma™ Supplier No. 61637020MG

Aprotinin, Bovine Lung, Crystalline, CAS 9087-70-1, is a competitive, reversible, heat- and acid-stable inhibitor of proteolytic and esterolytic activities.

Contaminants: endotoxin: ≤10 EU/mg

Specifications

Concentration 5500 KIU/mg, dry weight
Color White to Off-white
Target Proteins chymotrypsin
CAS 9087-70-1
Formula Weight 6512g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Crystalline Solid
Quantity 20 mg
Purity 95% by gel filtration