missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ SU5402

Catalog No. 5726302MG
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5726302MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5726302MG Supplier MilliporeSigma™ Supplier No. 5726302MG

SU5402, CAS 215543-92-3, is a cell-permeable, reversible, and ATP-competitive inhibitor of the tyrosine kinase activity of FGFR1 (IC50 = 10 to 20 μM in the presence of 1 mM ATP).

Cell Permeable

Specifications

Target Proteins Fibroblast growth factor receptor 1 (FGFR1)
Molecular Formula C17H16N2O3
Formula Weight 296.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Lyophilized
Quantity 2 mg
Purity 95% by HPLC