missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Staurosporine, Streptomyces sp.
Staurosporine, CAS 62996-74-, is a cell-permeable, potent, reversible, ATP-competitive inhibitor of protein kinases (IC50 = 7, 20, 1.3, 0.7, & 8.5 nM for PKA, CAMK, MLCK, PKC, & PKG, respectively).
$196.85 - $196.85
Specifications
| Color | White to Pale Yellow |
|---|---|
| Target Proteins | PKA |
| CAS | 62996-74-1 |
| Molecular Formula | C28H26N4O3 |
| Formula Weight | 466.5g/mol |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
56-939-7100UG
|
MilliporeSigma™
569397100UG |
100 μg |
Each for $196.85
|
|
|||||
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
| White to Pale Yellow | |
| 62996-74-1 | |
| 466.5g/mol | |
| Protect from light | |
| 99% by HPLC |
| PKA | |
| C28H26N4O3 | |
| DMSO or MeOH | |
| Solid |