missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Staurosporine, Streptomyces sp.

Catalog No. 569397100UG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
569397100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 569397100UG Supplier MilliporeSigma™ Supplier No. 569397100UG

Staurosporine, CAS 62996-74-, is a cell-permeable, potent, reversible, ATP-competitive inhibitor of protein kinases (IC50 = 7, 20, 1.3, 0.7, & 8.5 nM for PKA, CAMK, MLCK, PKC, & PKG, respectively).

  • Packaged Under Inert Gas
  • Cell Permeable

Specifications

Color White to Pale Yellow
Target Proteins PKA
CAS 62996-74-1
Molecular Formula C28H26N4O3
Formula Weight 466.5g/mol
Solubility DMSO or MeOH
Content And Storage Protect from light
Form Solid
Quantity 100 μg
Purity 99% by HPLC