missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ α-Amyloid Precursor Protein Modulator

Catalog No. 5657401MG
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5657401MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5657401MG Supplier MilliporeSigma™ Supplier No. 5657401MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A cell-permeable benzolactam derived PKC activator (K

A cell-permeable benzolactam derived PKC activator (Ki = 11.9nM for PKCα) that efficiently enhances non-amyloidogenic α-processing of amyloid precursor protein (APP) even at 100nM in human fibroblast AG06848. Displays a marked loss of tumor promoting activity.

Specifications

Color Orange
Description 95% by HPLC
Target Proteins PKC
CAS 497259-23-1
Molecular Formula C27H30F3N3O3
Formula Weight 501.5g/mol
Solubility DMSO or MeOH
Content And Storage Protect from light
Form Solid
Quantity 1 mg