missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Akt Inhibitor V, Triciribine
The Akt Inhibitor V, Triciribine, also referenced under CAS 35943-35-2, controls the biological activity of Akt.
$322.80 - $322.80
Specifications
| Target Proteins | Akt1, Akt 2, Akt3 |
|---|---|
| CAS | 35943-35-2 |
| Molecular Formula | C13H16N6O4 |
| Formula Weight | 320.3g/mol |
| Solubility | DMSO |
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
| Akt1, Akt 2, Akt3 | |
| C13H16N6O4 | |
| DMSO | |
| Solid |
| 35943-35-2 | |
| 320.3g/mol | |
| Protect from light | |
| 95% by HPLC |