missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ γ-Secretase Inhibitor XX
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
Specifications
| Color | White |
| Product Type | Inhibitor |
| Target Proteins | γ-secretase |
| CAS | 209984-56-5 |
| Molecular Formula | C26H23F2N3O3 |
| Formula Weight | 463.4g/mol |
| Solubility | DMSO |
| Content And Storage | Protect from light |
| Form | Solid |
| Quantity | 500 μg |
| Show More |