missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ InSolution™ γ-Secretase Inhibitor IX
Description
Packaged Under Inert Gas
Specifications
Specifications
| Color | Clear Colorless |
| Formulation | A 25 mM (5mg/462μl or 10mg/924μl) solution of γ-Secretase Inhibitor IX in DMSO. |
| Target Proteins | Aβtotal |
| Molecular Formula | C23H26F2N2O4 |
| Formula Weight | 432.5g/mol |
| Content And Storage | Hygroscopic, Protect from light |
| Form | Liquid |
| Quantity | 5 mg |
| Purity | 95% by HPLC |