missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Rho Kinase Inhibitor

Catalog No. 5555505MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5555505MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5555505MG Supplier MilliporeSigma™ Supplier No. 5555505MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Rho kinase inhibitor, CAS 872543-07-6, is a cell-permeable, highly specific, reversible, potent, and ATP-competitive inhibitor of Rho-associated kinase (ROCK; Ki = 1.6 nM).

  • Packaged Under Inert Gas
  • Cell Permeable

Specifications

Color White
Target Proteins ROCK
Molecular Formula C16H21N3O2S·2HCl
Formula Weight 392.3g/mol
Solubility H2O or MeOH
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 5 mg
Purity 95% by HPLC