missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ PP2

Catalog No. 5295731MG
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5295731MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5295731MG Supplier MilliporeSigma™ Supplier No. 5295731MG

PP2, CAS 172889-27-9, is a potent, reversible, ATP-competitive, inhibitor of the Src family of protein tyrosine kinases (IC50 = 4, 5, 5, & 100 nM for p56lck, p59fynT, Hck, & Src, respectively).

Packaged Under Inert Gas

Specifications

Color Off-white
Target Proteins p56lck
CAS 172889-27-9
Molecular Formula C15H16ClN5
Formula Weight 301.8g/mol
Solubility DMSO
Form Solid
Quantity 1 mg
Purity 95% by HPLC