missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Phosphatase Inhibitor Cocktail Set III

Phosphatase Inhibitor Cocktail Set III is a ready to use cocktail of four phosphatase inhibitors for broad-spectrum inhibition of both serine/threonine and protein tyrosine phosphatases.

$972.56 - $972.56

Specifications

Formulation In H2O.
Target Proteins Serine/threonine and protein tyrosine phosphatases
Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
52-462-71SET
View Documents
MilliporeSigma™
5246271SET
1 Set
Each for $972.56
Only null left
 
Description

Description

A cocktail of four phosphatase inhibitors for broad-spectrum inhibition of both serine/threonine and protein tyrosine phosphatases. Available as a 1 ml vial or as a set of five 1 ml vials. Each vial contains 1 ml of aqueous solution with the following components: 50mm Sodium Fluoride, 10mm β-Glycerophosphate (Cat. No. 35675), 10mm Sodium Pyrophosphate Decahydrate, 1mm Sodium Orthovanadate.
Specifications

Specifications

In H2O.
Serine/threonine and protein tyrosine phosphatases
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'