missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Phosphatase Inhibitor Cocktail Set III

Catalog No. 5246271SET
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5246271SET 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5246271SET Supplier MilliporeSigma™ Supplier No. 5246271SET

Phosphatase Inhibitor Cocktail Set III is a ready to use cocktail of four phosphatase inhibitors for broad-spectrum inhibition of both serine/threonine and protein tyrosine phosphatases.

A cocktail of four phosphatase inhibitors for broad-spectrum inhibition of both serine/threonine and protein tyrosine phosphatases. Available as a 1mL vial or as a set of five 1mL vials. Each vial contains 1mL of aqueous solution with the following components: 50mm Sodium Fluoride, 10mm β-Glycerophosphate (Cat. No. 35675), 10mm Sodium Pyrophosphate Decahydrate, 1mm Sodium Orthovanadate.

Specifications

Formulation In H2O.
Target Proteins Serine/threonine and protein tyrosine phosphatases
Form Liquid
Quantity 1 Set