missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Phosphatase Inhibitor Cocktail Set IV

Catalog No. 5246281ml
Change view
Click to view available options
Quantity:
1 Set
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5246281ML 1 mL
5246281SET 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5246281ML Supplier MilliporeSigma™ Supplier No. 5246281ML
Only null left

A cocktail of three phosphatase inhibitors for the inhibition of both serine/threonine and alkaline phosphatases.

A cocktail of three phosphatase inhibitors for the inhibition of both serine/threonine and alkaline phosphatases. Available as a 1mL vial or as a set of five 1mL vials. Each vial contains a 1mL solution with the following components: 500μM Cantharidin (Cat. No. 210155), 2.5mm (-)- p -Bromotetramisole oxalate, and 1μM Calyculin A, Discodermia calyx (Cat. No. 208851).

Specifications

Formulation In 95% DMSO.
Target Proteins Serine/threonine and alkaline phosphatases
Content And Storage Protect from light
Form Liquid
Quantity 1 mL