missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Akt Inhibitor VII, TAT-Akt-in
Description
- Packaged Under Inert Gas
- Peptide Sequence: H-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-Ala-Val-Thr-Asp-His-Pro-Asp-Arg-Leu-Trp-Ala-Trp-Glu-Lys-Phe-OH
- Cell Permeable
Specifications
Specifications
| Color | White |
| Formulation | Supplied as an trifluoroacetate salt. |
| Description | 95% by HPLC |
| Product Type | Inhibitor |
| Target Proteins | Akt |
| Molecular Formula | C152H239N55O36 |
| Formula Weight | 3412.9g/mol |
| Solubility | H2O |
| Content And Storage | Hygroscopic |
| Form | Solid |
| Show More |