missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ NF-κB SN50, Cell-Permeable Inhibitor Peptide
NF-κB SN50, CAS 213546-43-3, is a cell-permeable peptide that contains the NLS sequence of NF-κB p50 linked to the hydrophobic region of K-FGF. Inhibits translocation of the NF-κB into the nucleus.
$486.51 - $486.51
Specifications
| Formulation | Supplied as a trifluoroacetate salt. |
|---|---|
| Target Proteins | translocation of the NF-κB active complex into the nucleus |
| Molecular Formula | C129H230N36O29S |
| Formula Weight | 2781.5g/mol |
| Solubility | H2O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
48-148-01MG
|
MilliporeSigma™
4814801MG |
1 mg |
Each for $486.51
|
|
|||||
Description
- Peptide Sequence: H-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Val-Gln-Arg-Lys-Arg-Gln-Lys-Leu-Met-Pro-OH
- Cell Permeable
Specifications
| Supplied as a trifluoroacetate salt. | |
| C129H230N36O29S | |
| H2O | |
| Lyophilized |
| translocation of the NF-κB active complex into the nucleus | |
| 2781.5g/mol | |
| Hygroscopic | |
| 97% by HPLC |