missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ MG-132

Catalog No. p-7060104
Change view
Click to view available options
Quantity:
1 mg
5 mg
Catalog No. Quantity
4747911MG 1 mg
4747915MG 5 mg
2 options

Catalog No. 4747911MG

Supplier: MilliporeSigma™ 4747911MG

Only null left

InSolution™ MG-132, CAS 133407-82-6, is provided as a 10 mM solution in DMSO. Acts as a potent, reversible proteasome inhibitor (Ki = 4 nM).

  • Packaged Under Inert Gas
  • Cell Permeable

Specifications

Formulation A 10 mM (1mg/210μl) or (5mg/1050μl) solution ofmg-132 in DMSO.
Target Proteins proteasome
CAS 133407-82-6
Molecular Formula C26H41N3O5
Formula Weight 475.6g/mol
Content And Storage Protect from light
Form Liquid
Quantity 1 mg
Purity 98% by HPLC