missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ InSolution™ MG-132

Catalog No. 4747911MG
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4747911MG 1 mg
4747915MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4747911MG Supplier MilliporeSigma™ Supplier No. 4747911MG

InSolution™ MG-132, CAS 133407-82-6, is provided as a 10 mM solution in DMSO. Acts as a potent, reversible proteasome inhibitor (Ki = 4 nM).

  • Packaged Under Inert Gas
  • Cell Permeable

Specifications

Formulation A 10 mM (1mg/210μl) or (5mg/1050μl) solution ofmg-132 in DMSO.
Target Proteins proteasome
CAS 133407-82-6
Molecular Formula C26H41N3O5
Formula Weight 475.6g/mol
Content And Storage Protect from light
Form Liquid
Quantity 1 mg
Purity 98% by HPLC