missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ MG-132

Catalog No. 4747901MG
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4747901MG 1 mg
502306256 5 mg
47479010MG 10 mg
47479020MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 4747901MG Supplier MilliporeSigma™ Supplier No. 4747901MG

A cell-permeable, potent, reversible proteasome inhibitor (Ki = 4 nM).

  • Peptide Sequence: Z-Leu-Leu-Leu-CHO
  • Cell Permeable

Specifications

Color White
Target Proteins proteasome
CAS 133407-82-6
Molecular Formula C26H41N3O5
Formula Weight 475.6g/mol
Solubility DMSO or EtOH
Form Solid
Quantity 1 mg
Purity 98% by HPLC