missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ AG 490

Catalog No. 6584015MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6584015MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6584015MG Supplier MilliporeSigma™ Supplier No. 6584015MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A cell-permeable, reversible, substrate competitive, and potent inhibitor of epidermal growth factor receptor kinase autophosphorylation (IC50 = 100 nM).

Cell Permeable

Specifications

Color Yellow
Description 98% by HPLC
Product Type Inhibitor
Target Proteins epidermal growth factor receptor kinase autophosphorylation
CAS 133550-30-8
Molecular Formula C17H14N2O3
Formula Weight 294.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Show More Show Less