missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ AG 1024

Catalog No. 1217671MG
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1217671MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1217671MG Supplier MilliporeSigma™ Supplier No. 1217671MG

A cell-permeable, reversible, substrate competitive, and specific inhibitor of insulin-like growth factor-1 (IGF-1) and insulin receptor tyrosine kinase activity.

  • Packaged Under Inert Gas
  • Cell Permeable

Specifications

Color Off-white to Yellow
Description 98% by HPLC
Target Proteins IGF-1
CAS 65678-07-1
Molecular Formula C14H13BrN2O
Formula Weight 305.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg