missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ KT5720

Catalog No. 420320100UG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
420320100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 420320100UG Supplier MilliporeSigma™ Supplier No. 420320100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A potent, specific, cell-permeable, reversible and ATP-competitive inhibitor of protein kinase A (Ki = 56 nM) that is prepared by a chemical modification of K-252a.

Cell Permeable

Specifications

Color White
Target Proteins PKA
CAS 108068-98-0
Molecular Formula C32H31N3O5
Formula Weight 537.6g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 100 μg
Purity 95% by HPLC