missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ ACE2 Inhibitor, MLN-4760

Catalog No. 5306160001
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5306160001 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5306160001 Supplier MilliporeSigma™ Supplier No. 5.30616.0001

A cell-permeable, highly potent inhibitor of angiotensin converting enzyme 2 (ACE2; IC50 = 440 pM).

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach

Specifications

Color White
Description 97% by HPLC
Target Proteins soluble human ACE2
CAS 305335-31-3
Molecular Formula C19H23Cl2N3O4
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 2 mg