missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ AMPK Inhibitor, Compound C

Catalog No. 1712601mg
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1712601MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1712601MG Supplier MilliporeSigma™ Supplier No. 1712601MG
Only null left

AMPK Inhibitor, Compound C, CAS 866405-64-3, is a cell-permeable compound that inhibits KDR/VEGFR2, ALK2/BMPR-I, and AMPK kinase activities (IC50 = 25.1, 148, and 234.6 nM, respectively).

  • Packaged Under Inert Gas
  • Cell Permeable

Specifications

Color Light Yellow
Target Proteins AMPK
CAS 866405-64-3
Molecular Formula C24H25N5O
Formula Weight 399.5g/mol
Solubility DMSO, 100 mM HCl, or MeOH
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Purity 95% by HPLC