missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Chemical Name or Material | Cal-Ex™ II Fixative/Decalcifier |
|---|---|
| Physical Form | Liquid |
| CAS | 50-00-0 , 67-56-1 , 7732-18-5 |
| Packaging | Poly Bottle |
| Color | Colorless |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
CS5111D
|
Fisher Chemical
CS511 1 GH |
1 L |
Each for $130.19
Case of 6 Each for $781.14
|
|
|||||
|
CS5114D
|
Fisher Chemical
CS511 4 GH |
4 L |
Each for $217.81
Case of 4 Each for $871.24
|
|
|||||
Description
For simultaneous fixation and decalcification of tissue specimens prior to staining.Safety and Handling
DOTInformation : DOT Class 8, : Corrosive