missing translation for 'onlineSavingsMsg'
Learn More

Cal-Ex™ II Fixative/Decalcifier, Fisher Chemical™

Catalog No. CS5114D
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CS5114D 4 L
CS5111D 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CS5114D Supplier Fisher Chemical Supplier No. CS511 4 GH
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For simultaneous fixation and decalcification of tissue specimens prior to staining.

Specifications

Physical Form Liquid
Chemical Name or Material Cal-Ex™ II Fixative/Decalcifier
CAS 50-00-0, 67-56-1, 7732-18-5
Packaging Poly Bottle
Color Colorless
pH 2.0
Quantity 4 L
MDL Number MFCD00211744