missing translation for 'onlineSavingsMsg'
Learn More

Caffeine Solution, 1.0 mg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111017866
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017866 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017866 Supplier Merck Emd Millipore Supplier No. C60351ML
Only null left
Pharmaceutical (small molecule)

Specifications

Packaging Ampule
CAS 58-08-2
Flash Point 11°C
MDL Number MFCD00005758
Type Analytical Standard
Quantity 1 mL
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 1.0 mg/mL in methanol