missing translation for 'onlineSavingsMsg'
Learn More

Caffeine Melting Point Standard Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014745
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014745 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014745 Supplier Merck Emd Millipore Supplier No. PHR10951G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Caffeine is mainly used as a diuretic and as a stimulant to central nervous system, heart rate, and respiration. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 58-08-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C8H10N4O2
MDL Number MFCD00005758
Melting Point ∼237°C; 234°C to 236.5°C
Synonym Caffeine; 1,3,7-Trimethylxanthine
Type Certified Reference Material
Quantity 1 g
Formula Weight 194.19
Grade Certified Reference Material
Show More Show Less