missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Caerulein, Sulfated
Description
Stimulates smooth muscle and increases digestive secretions
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Purification Method | HPLC |
| Common Name | Ceruletide |
| Content And Storage | -30°C to -10°C |
| Format | Powder |
| For Use With (Application) | Stimulates smooth muscle and increases digestive secretions |
| Molecular Weight (g/mol) | 1352.40 |
| Product Type | Caerulein, sulfated |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
| Sequence | Glp-Gln-Asp-Tyr(SO3H)-Thr-Gly-Trp-Met-Asp-Phe-NH2 |
RUO – Research Use Only