missing translation for 'onlineSavingsMsg'
Learn More

Cadmium powder, -325 mesh, 99.5% (metals basis)

Catalog No. AA1189136
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1189136 500 g
AA1189109 10 g
AA1189122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1189136 Supplier Thermo Scientific Chemicals Supplier No. 01189136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cadmium powder, 99.5%, Cd, CAS Number-7440-43-9, ccris 112, unii-00bh33gngh, compounds, cadmium, elemental, cadmium and compounds, 00bh33gngh, kadmium, kadmium german, colloidal, hsdb 282, 500g, (metals basis), CHEBI:22977, Silver-White | CAS: 7440-43-9 | Cd | 112.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
PubChem CID 23973
ChEBI CHEBI:22977
IUPAC Name cadmium
SMILES [Cd]

Specifications

Color Silver-White
Quantity 500 g
Mesh Size −325 Mesh
Form Solid
UN Number UN3179
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Cadmium powder
Hazard Category H228-H330-H341-H350-H361fd-H372
Hazard Statement GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P210-P240-P241-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, TOXIC, INORGANIC, N.O.S.
EINECSNumber 231-152-8
RTECSNumber EU9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only