Learn More
Cadmium, plasma standard solution, Specpure™ Cd 1000μg/mL
CAS: 7440-43-9 | Cd | 112.41 g/mol
$139.99 - $139.99
Chemical Identifiers
| CAS | 7440-43-9 |
|---|---|
| Molecular Formula | Cd |
| Molecular Weight (g/mol) | 112.41 |
| MDL Number | MFCD00010914 |
| InChI Key | BDOSMKKIYDKNTQ-UHFFFAOYSA-N |
| PubChem CID | 23973 |
| ChEBI | CHEBI:22977 |
| IUPAC Name | cadmium |
| SMILES | [Cd] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-43-9 | |
| 112.41 | |
| BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
| CHEBI:22977 | |
| [Cd] |
| Cd | |
| MFCD00010914 | |
| 23973 | |
| cadmium |
Specifications
| 7440-43-9 , 7732-18-5 | |
| Liquid | |
| MFCD00010914 | |
| BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
| cadmium | |
| 100 mL | |
| CHEBI:22977 | |
| Cadmium |
| 23 hPa (17mm Hg) at 20°C | |
| Cd | |
| Colorless | |
| [Cd] | |
| 112.41 | |
| 23973 | |
| Nitric acid: 5.0%; Cadmium: 0.1%; Water: 94.9% |
Safety and Handling
When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
P201-P202-P234-P264b-P281-P302+P352-P305+P351+P338-P308+P313-P310-P332+P313-P362-P390-P501c
H290-H315-H318-H350
DOTInformation : Hazard Class: 8, Packing Group: III
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only