CAS RN 7440-43-9
Cadmium wire, 0.5mm (0.02in) dia, Puratronic™, 99.998% (metals basis)
CAS: 7440-43-9 Molecular Formula: Cd Molecular Weight (g/mol): 112.41 MDL Number: MFCD00010914 InChI Key: BDOSMKKIYDKNTQ-UHFFFAOYSA-N Synonym: kadmium,compounds,colloidal,cadmium, elemental,kadmium german,unii-00bh33gngh,ccris 112,hsdb 282,00bh33gngh,cadmium and compounds PubChem CID: 23973 ChEBI: CHEBI:22977 IUPAC Name: cadmium SMILES: [Cd]
Cadmium foil, 2.0mm (0.08in) thick, Puratronic™, 99.998% (metals basis)
CAS: 7440-43-9 Molecular Formula: Cd Molecular Weight (g/mol): 112.41 MDL Number: MFCD00010914 InChI Key: BDOSMKKIYDKNTQ-UHFFFAOYSA-N Synonym: kadmium,compounds,colloidal,cadmium, elemental,kadmium german,unii-00bh33gngh,ccris 112,hsdb 282,00bh33gngh,cadmium and compounds PubChem CID: 23973 ChEBI: CHEBI:22977 SMILES: [Cd]
Cadmium shot, 1-3mm (0.04-0.1in), Puratronic™, 99.9997% (metals basis)
CAS: 7440-43-9 Molecular Formula: Cd Molecular Weight (g/mol): 112.41 MDL Number: MFCD00010914 InChI Key: BDOSMKKIYDKNTQ-UHFFFAOYSA-N Synonym: kadmium,compounds,colloidal,cadmium, elemental,kadmium german,unii-00bh33gngh,ccris 112,hsdb 282,00bh33gngh,cadmium and compounds PubChem CID: 23973 ChEBI: CHEBI:22977 SMILES: [Cd]
Cadmium foil, 1.0mm (0.04in) thick, Puratronic™, 99.998% (metals basis)
CAS: 7440-43-9 Molecular Formula: Cd Molecular Weight (g/mol): 112.41 MDL Number: MFCD00010914 InChI Key: BDOSMKKIYDKNTQ-UHFFFAOYSA-N Synonym: kadmium,compounds,colloidal,cadmium, elemental,kadmium german,unii-00bh33gngh,ccris 112,hsdb 282,00bh33gngh,cadmium and compounds PubChem CID: 23973 ChEBI: CHEBI:22977 IUPAC Name: cadmium(2+) ion SMILES: [Cd]
Cadmium rod, 8mm (0.3in) dia, Puratronic™, 99.999% (metals basis)
CAS: 7440-43-9 Molecular Formula: Cd Molecular Weight (g/mol): 112.41 MDL Number: MFCD00010914 InChI Key: BDOSMKKIYDKNTQ-UHFFFAOYSA-N Synonym: kadmium,compounds,colloidal,cadmium, elemental,kadmium german,unii-00bh33gngh,ccris 112,hsdb 282,00bh33gngh,cadmium and compounds PubChem CID: 23973 ChEBI: CHEBI:22977 IUPAC Name: cadmium SMILES: [Cd]
Cadmium foil, 0.5mm (0.02in) thick, Puratronic™, 99.9975% (metals basis)
CAS: 7440-43-9 Molecular Formula: Cd Molecular Weight (g/mol): 112.41 MDL Number: MFCD00010914 InChI Key: BDOSMKKIYDKNTQ-UHFFFAOYSA-N Synonym: kadmium,compounds,colloidal,cadmium, elemental,kadmium german,unii-00bh33gngh,ccris 112,hsdb 282,00bh33gngh,cadmium and compounds PubChem CID: 23973 ChEBI: CHEBI:22977 SMILES: [Cd]
Cadmium foil, 0.25mm (0.01in) thick, Puratronic™, 99.9975% (metals basis)
CAS: 7440-43-9 Molecular Formula: Cd Molecular Weight (g/mol): 112.41 MDL Number: MFCD00010914 InChI Key: BDOSMKKIYDKNTQ-UHFFFAOYSA-N Synonym: kadmium,compounds,colloidal,cadmium, elemental,kadmium german,unii-00bh33gngh,ccris 112,hsdb 282,00bh33gngh,cadmium and compounds PubChem CID: 23973 ChEBI: CHEBI:22977 SMILES: [Cd]
Cadmium wire, 0.3mm (0.013 in.) dia., Puratronic™, 99.999% (metals basis), Thermo Scientific™
CAS: 7440-43-9 Molecular Formula: Cd Molecular Weight (g/mol): 112.41 MDL Number: MFCD00010914 InChI Key: BDOSMKKIYDKNTQ-UHFFFAOYSA-N Synonym: kadmium,compounds,colloidal,cadmium, elemental,kadmium german,unii-00bh33gngh,ccris 112,hsdb 282,00bh33gngh,cadmium and compounds PubChem CID: 23973 ChEBI: CHEBI:22977 IUPAC Name: cadmium SMILES: [Cd]
Cadmium, plasma standard solution, Specpure™ Cd 1000μg/mL
CAS: 7440-43-9 Molecular Formula: Cd Molecular Weight (g/mol): 112.41 MDL Number: MFCD00010914 InChI Key: BDOSMKKIYDKNTQ-UHFFFAOYSA-N PubChem CID: 23973 ChEBI: CHEBI:22977 IUPAC Name: cadmium SMILES: [Cd]
Cadmium shot, 5cm (2 in.) dia., 99.9% (metals basis), Thermo Scientific™
CAS: 7440-43-9 Molecular Formula: Cd Molecular Weight (g/mol): 112.41 MDL Number: MFCD00010914 InChI Key: BDOSMKKIYDKNTQ-UHFFFAOYSA-N Synonym: kadmium,compounds,colloidal,cadmium, elemental,kadmium german,unii-00bh33gngh,ccris 112,hsdb 282,00bh33gngh,cadmium and compounds PubChem CID: 23973 ChEBI: CHEBI:22977 IUPAC Name: cadmium SMILES: [Cd]
Cadmium foil, 0.05mm (0.002in) thick, Puratronic™, 99.999% (metals basis)
CAS: 7440-43-9 Molecular Formula: Cd Molecular Weight (g/mol): 112.41 MDL Number: MFCD00010914 InChI Key: BDOSMKKIYDKNTQ-UHFFFAOYSA-N Synonym: kadmium,compounds,colloidal,cadmium, elemental,kadmium german,unii-00bh33gngh,ccris 112,hsdb 282,00bh33gngh,cadmium and compounds PubChem CID: 23973 ChEBI: CHEBI:22977 IUPAC Name: cadmium SMILES: [Cd]
Cadmium powder, -325 mesh, 99.5% (metals basis)
CAS: 7440-43-9 Molecular Formula: Cd Molecular Weight (g/mol): 112.41 MDL Number: MFCD00010914 InChI Key: BDOSMKKIYDKNTQ-UHFFFAOYSA-N Synonym: kadmium,compounds,colloidal,cadmium, elemental,kadmium german,unii-00bh33gngh,ccris 112,hsdb 282,00bh33gngh,cadmium and compounds PubChem CID: 23973 ChEBI: CHEBI:22977 IUPAC Name: cadmium SMILES: [Cd]
Cadmium foil, 0.1mm (0.004in) thick, Puratronic™, 99.9975% (metals basis)
CAS: 7440-43-9 Molecular Formula: Cd Molecular Weight (g/mol): 112.41 MDL Number: MFCD00010914 InChI Key: BDOSMKKIYDKNTQ-UHFFFAOYSA-N Synonym: kadmium,compounds,colloidal,cadmium, elemental,kadmium german,unii-00bh33gngh,ccris 112,hsdb 282,00bh33gngh,cadmium and compounds PubChem CID: 23973 ChEBI: CHEBI:22977 SMILES: [Cd]
Cadmium shot, tear drop, -5+20 mesh, 99.95% (metals basis)
CAS: 7440-43-9 Molecular Formula: Cd Molecular Weight (g/mol): 112.41 MDL Number: MFCD00010914 InChI Key: BDOSMKKIYDKNTQ-UHFFFAOYSA-N Synonym: kadmium,compounds,colloidal,cadmium, elemental,kadmium german,unii-00bh33gngh,ccris 112,hsdb 282,00bh33gngh,cadmium and compounds PubChem CID: 23973 ChEBI: CHEBI:22977 IUPAC Name: cadmium SMILES: [Cd]
Cadmium rod, 12.7mm (0.5in) dia, 99.99+% (metals basis)
CAS: 7440-43-9 Molecular Formula: Cd Molecular Weight (g/mol): 112.41 MDL Number: MFCD00010914 InChI Key: BDOSMKKIYDKNTQ-UHFFFAOYSA-N Synonym: kadmium,compounds,colloidal,cadmium, elemental,kadmium german,unii-00bh33gngh,ccris 112,hsdb 282,00bh33gngh,cadmium and compounds PubChem CID: 23973 ChEBI: CHEBI:22977 IUPAC Name: cadmium SMILES: [Cd]