missing translation for 'onlineSavingsMsg'
Learn More

Cadmium oxide, 99%, pure

Catalog No. AC223792500
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC223792500 250 g
AC223790010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC223792500 Supplier Thermo Scientific Chemicals Supplier No. 223792500
Only null left

CAS: 1306-19-0 | CdO | 128.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1306-19-0
Molecular Formula CdO
Molecular Weight (g/mol) 128.41
MDL Number MFCD00010921
InChI Key CFEAAQFZALKQPA-UHFFFAOYSA-N
Synonym cadmium oxide, cadmium monoxide, cadmium fume, aska-rid, kadmu tlenek polish, caswell no. 136aa, ccris 115, epa pesticide chemical code 236200, dsstox_cid_4715
PubChem CID 14782
IUPAC Name oxocadmium
SMILES [O--].[Cd++]

Specifications

Boiling Point 1385.0°C
Quantity 250 g
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula CdO
Merck Index 15,1628
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diluted acids and alkalies
Formula Weight 128.41
Percent Purity 99%
Grade Pure
Chemical Name or Material Cadmium oxide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.
Fatal if inhaled.
May cause cancer.
Suspected
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF INHALED: Remove to fresh air and keep at rest in a
EINECSNumber 215-146-2

RUO – Research Use Only