missing translation for 'onlineSavingsMsg'
Learn More

Cadmium iodide, 98%

Catalog No. AAA1807914
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1807914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1807914 Supplier Thermo Scientific Chemicals Supplier No. A1807914
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 7790-80-9 | CdI2 | 366.223 g/mol

It finds it use in photography, process engraving, lithography, analytical chemistry, and electroplating; Also used as in lubricants, phosphors. Used for cadmium electrode position and in photoconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it use in photography, process engraving, lithography, analytical chemistry, and electroplating; Also used as in lubricants, phosphors. Used for cadmium electrode position and in photoconductors.

Solubility
Very soluble in water, slightly soluble in acetone, ethanol and diethyl ether.

Notes
Light Sensitive & Hygroscopic. Store away from oxidizing agents, light and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 7790-80-9
Molecular Formula CdI2
Molecular Weight (g/mol) 366.223
MDL Number MFCD00010920
InChI Key OKIIEJOIXGHUKX-UHFFFAOYSA-L
Synonym cadmium iodide, cadmium diiodide, unii-2f2upu4kcw, cadmium 2+ diiodide, 2f2upu4kcw, acmc-20aljc, cadmium 2+ ion diiodide, cadmium iodide trace metals basis 25g
PubChem CID 24635
IUPAC Name cadmium(2+);diiodide
SMILES [Cd+2].[I-].[I-]

Specifications

Boiling Point 787°C
Melting Point 387°C to 389°C
Quantity 25 g
UN Number UN2570
Merck Index 14,1621
Solubility Information Very soluble in water,slightly soluble in acetone,ethanol and diethyl ether.
Formula Weight 366.21
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Density 5.67
Chemical Name or Material Cadmium iodide
Show More Show Less
Hazard Category H301+H331-H351-H373
Hazard Statement GHS H Statement
H301-H331-H351-H373
Toxic if swallowed.
Toxic if inhaled.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
EINECSNumber 232-223-6
RTECSNumber EV1290000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only