missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium ICP Standard, 10,000 ppm Cd in 5% HNO3, Ricca Chemical
$528.90 - $1049.47
Chemical Identifiers
| CAS | 7440-43-9 |
|---|---|
| Molecular Formula | Cd |
| Molecular Weight (g/mol) | 112.41 |
| MDL Number | MFCD00010914 |
| InChI Key | BDOSMKKIYDKNTQ-UHFFFAOYSA-N |
| PubChem CID | 23973 |
| ChEBI | CHEBI:22977 |
| IUPAC Name | cadmium |
| SMILES | [Cd] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PCD10KN-100
|
Ricca Chemical Company
PCD10KN100 |
100 mL | Natural LDPE Bottle |
Each for $528.90
|
|
||||
|
PCD10KN-500
|
Ricca Chemical Company
PCD10KN500 |
500 mL | Natural Poly Bottle |
Each for $1,049.47
|
|
||||
Chemical Identifiers
| 7440-43-9 | |
| 112.41 | |
| BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
| CHEBI:22977 | |
| [Cd] |
| Cd | |
| MFCD00010914 | |
| 23973 | |
| cadmium |
Specifications
| Natural LDPE Bottle | |
| 10,000ppm Cd | |
| 91.0,10.24,0.91 | |
| Liquid | |
| MFCD00010914 | |
| < 2 | |
| BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
| cadmium | |
| 100 mL | |
| CHEBI:22977 |