missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Chloride, Hydrated, Crystal, 79.5-81.0%, Spectrum™ Chemical

Catalog No. 18609408
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609408 2.5 kg Amber Glass Bottle
18609405 25 g Amber Glass Bottle
18609406 125 g Amber Glass Bottle
18609407 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609408 Supplier Spectrum Chemical Mfg Cor Supplier No. C10262.5KG
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7790-78-5

Specifications

CAS Max % 1
Physical Form Crystals or Needles
Quantity 2.5 kg
Percent Purity 79.5 to 81.0%
Grade Reagent
Packaging Amber Glass Bottle
Chemical Name or Material Cadmium Chloride, Hydrated, Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.