missing translation for 'onlineSavingsMsg'
Learn More

Cadmium AA Standard, 1000 ppm Cd in 3% HNO3, Ricca Chemical

$124.23 - $124.23

Chemical Identifiers

CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
PubChem CID 23973
ChEBI CHEBI:22977
IUPAC Name cadmium
SMILES [Cd]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
ACD1KN-100
View Documents
Ricca Chemical Company
ACD1KN100
100 mL Natural LDPE Bottle
Each for $124.23
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7440-43-9
112.41
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
CHEBI:22977
[Cd]
Cd
MFCD00010914
23973
cadmium

Specifications

Natural LDPE Bottle
1,000ppm Cd
97.0,4.74,0.1
Liquid
MFCD00010914
< 2
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
cadmium
100 mL
CHEBI:22977
7440-43-9 , 7697-37-2 , 7440-43-9
93.0,4.56,0.09
1 mL = 1 mg Cd
Cd
Colorless
Approximately 0°C
[Cd]
112.41
23973
Cadmium AA Standard
Videos
SDS
Documents

Documents