missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CacyBP Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories Inc A302905AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
Calcyclin is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in stimulation of Ca2+-dependent insulin release, stimulation of prolactin secretion, and exocytosis. Chromosomal rearrangements and altered expression of this gene have been implicated in melanoma.Identifiants chimiques
| CacyBP | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| S100A6-binding protein; Siah-interacting protein (SIP); calcyclin-binding protein; growth-inhibiting gene 5 protein; hCacyBP | |
| Between 200 and 228 | |
| Primary | |
| 4° C | |
| CACYBP |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q9CXW3, Q9HB71 | |
| 12301, 27101 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
Spécifications
| CacyBP | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q9CXW3, Q9HB71 | |
| Cacybp | |
| Between 200 and 228 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| CACYBP | |
| S100A6-binding protein; Siah-interacting protein (SIP); calcyclin-binding protein; growth-inhibiting gene 5 protein; hCacyBP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12301, 27101 | |
| 4° C | |
| Liquid |