missing translation for 'onlineSavingsMsg'
Learn More

C7 - C30 Saturated Alkanes, Certified Reference Material, 1,000 μg/mL Each Component in Hexane, MilliporeSigma™ Supelco™

Catalog No. 111017206
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017206 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017206 Supplier Merck Emd Millipore Supplier No. 49451U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Quantitative standard suitable for use as a retention index marker probe.

Specifications

Packaging Ampule
Product Line TraceCERT
Concentration 1000 μg/mL each component in hexane
Quantity 1 mL
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) Decane; Docosane; Dodecane; Eicosane; Heneicosane; Heptacosane; Heptadecane; Hexacosane; Hexadecane; Heptane; Nonacosane; Nonadecane; Nonane; Octacosane; Octadecane; Octane; Pentacosane; Pentadecane; Tetracosane; Tetradecane; Triacontane; Tricosane; Tridecane; Undecane

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC