missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C19orf68 Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. PIPA563923
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PIPA563923 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIPA563923 Fournisseur Invitrogen™ Code fournisseur PA563923
Il en reste null

Rabbit Polyclonal Antibody

Immunogen sequence: RRLLSYCKGR DHGVLDALHV LEGLFRTDPE AKVKLVFVED QAVVETVFFL TSRTRALLRR FPRMLLVDRL Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 87%.

C19orf68 gene ontology annotations related to this gene include multicellular organism development; ubiquitin-dependent protein catabolic process.

Spécifications

Antigen C19orf68
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZSWIM9
Gene Accession No. Q86XI8
Gene Alias C19orf68; chromosome 19 open reading frame 68; uncharacterized protein C19orf68; Uncharacterized protein ZSWIM9; ZSWIM9
Gene Symbols ZSWIM9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C19orf68. Recombinant protein control fragment (Product #RP-101698).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374920
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats