Learn More
Invitrogen™ c-Rel Monoclonal Antibody (1RELAH5), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$2,150.00
Spécifications
| Antigen | c-Rel |
|---|---|
| Clone | 1RELAH5 |
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
14611137
|
Invitrogen™
14611137 |
2 mg |
chaque for $2,150.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Description: The 1RELAH5 monoclonal antibody reacts with mouse c-Rel. c-Rel is a 65 kDa hematopoietic lineage-restricted member of the NF-kB family of transcription factors. In the thymus, c-Rel is upregulated in regulatory T cells (Treg) when compared to other CD4+ thymocytes. c-Rel is believed to play a critical role in thymic Treg (nTreg) development, but is not necessary for TGF beta-mediated T reg (iTreg) differentiation. C-Rel is also involved in proliferation and survival in mature T cells. Applications Reported: This 1RELAH5 antibody has been reported for use in western blotting. Applications Tested: This 1RELAH5 antibody has been tested by western blot analysis of mouse splenocytes. This can be used at 1-5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
The REL gene encodes c-Rel, a transcription factor that is a member of the Rel/NFKB family, which also includes RELA., RELB (604758), NFKB1, and NFKB2. These proteins are related through a highly conserved N-terminal region termed the 'Rel domain, ' which is responsible for DNA binding, dimerization, nuclear localization, and binding to the NFKB inhibitor (Belguise and Sonenshein, 2007).Spécifications
| c-Rel | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 19696 | |
| Primary | |
| 4°C | |
| REL |
| 1RELAH5 | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P15307 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |