missing translation for 'onlineSavingsMsg'
Learn More

TECHREF™ C-700 Sample Cups/Cells, 31 mm, single open ended

Catalog No. 11022102
Change view
Click to view available options
Diameter (Metric):
31 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
11022102 31 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11022102 Supplier TECHREF™ Supplier No. C700

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed specifically for use with X-ray fluorescence analyzers. TECHREF™ Sample Cells are available for XRF analyzers for a variety of styles and manufacturers and are designed for ease of use.

  • Composed of two parts: closed-bottom cup, outer open ring to hold and seal the desired film
  • One fill line in the interior of cup for consistent sample size
  • Mated locking line to prevent leakage
  • Dimple on exterior floor for venting and reservoir for overflow
  • Fully disposable
  • Designed for X-Ray Fluorescence Analyzers for EDXRF and WDXRF analysis
  • Suitable for Oxford, Rigaku, Asoma, Olympus (Innov-X), XOS, Spectro-Ametek, PANalytical (Philips–certain models), Xcalibur, Shimadzu, Horiba X-5000 XRF analyzers

Specifications

For Use With (Equipment) Oxford, Rigaku, Asoma, Olympus (Innov-X), XOS, Spectro-Ametek, PANalytical (Philips: certain models)
Type Disposable Cell
Certifications/Compliance ISO 9001
Diameter (Metric) 31 mm
Height (Metric) 22 mm
Includes Closed-bottom cup, outer open ring
Material Polyethylene