missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ C 176
Description
C 176 is a mSTING antagonist. Covalently binds Cys91 of mouse but not human STING. Blocks activation-induced palmitoylation of STING and formation of STING clusters. Reduces CMA (Cat. No. 6677) -induced increase in serum IFNs in vivo and suppresses systemic inflammation in a mouse inflammatory disease model.
Spécifications
Spécifications
| CAS | 314054-00-7 |
| Quantity | 10 mg |
| Target | STING Antagonists |
| Molecular Formula | C11H7IN2O4 |
| Formula Weight | 808 |
| Purity | 97% |