missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BYSL Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556679
Description
Immunogen sequence: LDALVFHFLG FRTEKRELPV LWHQCLLTLV QRYKADLATD QKEALLELLR LQPHPQLSPE IRRELQSAVP RDVEDVPITV E Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.
Bystin is expressed as a 2-kb major transcript and a 3.6-kb minor transcript in SNG-M cells and in human trophoblastic teratocarcinoma HT-H cells. Protein binding assays determined that bystin binds directly to trophinin and tastin, and that binding is enhanced when cytokeratins 8 and 18 are present. Immunocytochemistry of HT-H cells showed that bystin colocalizes with trophinin, tastin, and the cytokeratins, suggesting that these molecules form a complex in trophectoderm cells at the time of implantation. Using immunohistochemistry it was determined that trophinin and bystin are found in the placenta from the sixth week of pregnancy. Both proteins were localized in the cytoplasm of the syncytiotrophoblast in the chorionic villi and in endometrial decidual cells at the uteroplacental interface. After week 10, the levels of trophinin, tastin, and bystin decreased and then disappeared from placental villi.Identifiants chimiques
| BYSL | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| BYSL | |
| by the ribosomal protein s6 gene, drosophila, homolog-like; Bys; BYSL; bysl {ECO:0000312; Bystin; bystin like; bystin-like; EMBL:AAH79030.1, ECO:0000312; Enp1; RGD:727959} | |
| BYSL | |
| Primary | |
| Antigen affinity chromatography |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O54825, Q13895 | |
| 53414, 705 | |
| Recombinant protein corresponding to Human BYSL. Recombinant protein control fragment (Product #RP-94698). | |
| Antibody |
Spécifications
| BYSL | |
| Polyclonal | |
| Unconjugated | |
| BYSL | |
| by the ribosomal protein s6 gene, drosophila, homolog-like; Bys; BYSL; bysl {ECO:0000312; Bystin; bystin like; bystin-like; EMBL:AAH79030.1, ECO:0000312; Enp1; RGD:727959} | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 53414, 705 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O54825, Q13895 | |
| BYSL | |
| Recombinant protein corresponding to Human BYSL. Recombinant protein control fragment (Product #RP-94698). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |