missing translation for 'onlineSavingsMsg'
Learn More

Butyryl Chloride 98.0+%, TCI America™

Catalog No. B0770500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0770500G 500 g
B077025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0770500G Supplier TCI America Supplier No. B0770500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 141-75-3
Molecular Formula C4H7ClO
Molecular Weight (g/mol) 106.55
MDL Number MFCD00000752
InChI Key DVECBJCOGJRVPX-UHFFFAOYSA-N
Synonym butyryl chloride, n-butyryl chloride, butyric acid chloride, butyroyl chloride, butyric chloride, butyrylchloride, unii-2xvm8e16ir, ccris 5989, n-butyric acid chloride, n-butanoyl chloride
PubChem CID 8855
IUPAC Name butanoyl chloride
SMILES CCCC(Cl)=O

Specifications

Color Yellow
Boiling Point 102°C
Quantity 500 g
UN Number 2353
Formula Weight 106.55
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Butyryl Chloride
EINECSNumber (2)-0632
TSCA Yes