missing translation for 'onlineSavingsMsg'
Learn More

Butyryl chloride, 98%

Catalog No. AAA1757530
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1757530 250 g
AAA175750B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1757530 Supplier Thermo Scientific Chemicals Supplier No. A1757530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 141-75-3 | C4H7ClO | 106.55 g/mol

Butyryl chloride is used as an intermediate for the synthesis of active pharmaceutical ingredients benzoyl peroxide, t-butyl perbenzoate, and benzophenone. It is also used as an acylation reagent. Further, it is used in the preparation of peroxides such as benzoyl peroxide, benzophenone and t-butyl perbenzoate. In addition to this, it is involved in the preparation of chromones and antiproliferative agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyryl chloride is used as an intermediate for the synthesis of active pharmaceutical ingredients benzoyl peroxide, t-butyl perbenzoate, and benzophenone. It is also used as an acylation reagent. Further, it is used in the preparation of peroxides such as benzoyl peroxide, benzophenone and t-butyl perbenzoate. In addition to this, it is involved in the preparation of chromones and antiproliferative agents.

Solubility
Miscible with ether.

Notes
Moisture sensitive. Incompatible with water, alcohols, strong bases and strong oxidizing agents

Chemical Identifiers

CAS 141-75-3
Molecular Formula C4H7ClO
Molecular Weight (g/mol) 106.55
MDL Number MFCD00000752
InChI Key DVECBJCOGJRVPX-UHFFFAOYSA-N
Synonym butyryl chloride, n-butyryl chloride, butyric acid chloride, butyroyl chloride, butyric chloride, butyrylchloride, unii-2xvm8e16ir, ccris 5989, n-butyric acid chloride, n-butanoyl chloride
PubChem CID 8855
IUPAC Name butanoyl chloride
SMILES CCCC(Cl)=O

Specifications

Melting Point -89°C
Density 1.026
Boiling Point 101°C to 102°C
Flash Point 18°C (64°F)
Odor Acrid
Refractive Index 1.412
Quantity 250 g
UN Number UN2353
Beilstein 605395
Sensitivity Moisture sensitive
Merck Index 14,1598
Solubility Information Miscible with ether.
Formula Weight 106.55
Percent Purity 98%
Chemical Name or Material Butyryl chloride
Show More Show Less
Hazard Category H225-H302-H314-H331
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTYRYL CHLORIDE
EINECSNumber 205-498-5
RTECSNumber EM0595000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only