missing translation for 'onlineSavingsMsg'
Learn More

Butyric Acid, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019490
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019490 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019490 Supplier Merck Emd Millipore Supplier No. 192155ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Butyric acid is a 4-carbon fatty acid which is a potent differentiation agent for cells and also possesses antiproliferative activity. In cultured mammalian cells it affects growth rate, morphology and gene expression. It also induces apoptosis.

Specifications

CAS 107-92-6
Vapor Pressure 0.43 mmHg (20°C)
Vapor Density 3.04 (vs. air)
Density 0.964 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3CH2CH2COOH
MDL Number MFCD00002814
Melting Point -6°C to +3°C
Boiling Point 162°C
Quantity 5 mL
Formula Weight 88.11
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less