missing translation for 'onlineSavingsMsg'
Learn More

Butyramide 98.0+%, TCI America™

Catalog No. B0809500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0809500G 500 g
B080925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0809500G Supplier TCI America Supplier No. B0809500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 541-35-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.122
MDL Number MFCD00041894
InChI Key DNSISZSEWVHGLH-UHFFFAOYSA-N
Synonym butyramide, n-butyramide, n-butylamide, butanimidic acid, butanoic acid, amide, unii-9j6or937vr, butyroamide, n-c3h7c o nh2, butyrylamide, butyramid
PubChem CID 10927
ChEBI CHEBI:50724
IUPAC Name butanamide
SMILES CCCC(=O)N

Specifications

Melting Point 115°C
Color White
Boiling Point 216°C
Quantity 500 g
Formula Weight 87.12
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material Butyramide