missing translation for 'onlineSavingsMsg'
Learn More

Butyraldoxime 95.0+%, TCI America™

Catalog No. B0753500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0753500ML 500 mL
B075325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0753500ML Supplier TCI America Supplier No. B0753500ML

Chemical Identifiers

CAS 110-69-0
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.122
MDL Number MFCD00013943
InChI Key KGGVGTQEGGOZRN-SNAWJCMRSA-N
Synonym butyraldehyde oxime, butanal oxime, n-butyraldoxime, butylaldoxime, butanal, oxime, butyraldoxime, exkin 1, troykyd anti-skin bto, usaf am-6, m-butyraldehyde oxime
PubChem CID 5324278
IUPAC Name (NE)-N-butylidenehydroxylamine
SMILES CCCC=NO

Specifications

Color Yellow
Boiling Point 152°C
Quantity 500 mL
UN Number 2840
Formula Weight 87.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyraldoxime
EINECSNumber (2)-0504
RTECSNumber ES3500000
TSCA Yes